Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diindolylmethane, 96%, Thermo Scientific™

Catalog No. AC459220250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459220250 25 g
AC459220050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459220250 Supplier Thermo Scientific Chemicals Supplier No. 459220250

Chemical Identifiers

CAS 1968-05-4
Molecular Formula C17H14N2
Molecular Weight (g/mol) 246.31

Specifications

Melting Point 164.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 25 g
Synonym heptane, 3-bromo, 3-heptyl bromide, acmc-1bscs
InChI Key MLHXKYLLJRLHGH-UHFFFAOYNA-N
SMILES CCCCC(CC)Br
IUPAC Name 3-bromoheptane
PubChem CID 16091
Formula Weight 246.31
Percent Purity 96%
Chemical Name or Material 3,3'-Diindolylmethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.