Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1,2-epoxybutane, 95%

Catalog No. aal0695106
Click to view available options
Quantity:
5 g
25 g

CAS: 2245-30-9 | C6H12O | 100.161 g/mol

3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-30-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00051590
InChI Key HEAYDCIZOFDHRM-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1,2-epoxybutane, tert-butyloxirane, tert-butylethylene oxide, 3,3-dimethylbutene oxide, oxirane, 1,1-dimethylethyl, 3,3-dimethyl-1-butene oxide, 1,2-epoxy-3,3-dimethylbutane, tert.-butyloxirane, tert-butyl oxirane, butane, 1,2-epoxy-3,3-dimethyl
PubChem CID 92174
IUPAC Name 2-tert-butyloxirane
SMILES CC(C)(C)C1CO1

Specifications

Density 0.822
Boiling Point 95°C to 96°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.401
Quantity 5 g
UN Number UN1993
Beilstein 102478
Formula Weight 100.16
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1,2-epoxybutane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 218-831-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.