Learn More
3,3-Dimethyl-1-butanol, 97%
Description
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | -60°C |
| Density | 0.81 |
| Boiling Point | 142°C to 143°C |
| Flash Point | 56°C (133°F) |
| Odor | Alcohol-like |
| Refractive Index | 1.414 |
| Linear Formula | (CH3)3CCH2CH2OH |
| Quantity | 10 g |
| UN Number | UN1987 |
| Solubility Information | Slightly soluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S. |
| EINECSNumber | 210-872-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.