Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-1-butene 96.0+%, TCI America™
SDP

Catalog No. D1163100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1163100ML 100 mL
D116325ML 25 mL
D1163500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1163100ML Supplier TCI America Supplier No. D1163100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 558-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008853
InChI Key PKXHXOTZMFCXSH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk
PubChem CID 11210
IUPAC Name 3,3-dimethylbut-1-ene
SMILES CC(C)(C)C=C

Specifications

Color Colorless
Boiling Point 41°C
Quantity 100 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.