Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,3-Dimethylacrylic acid, 98%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Chemical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chemical Intermediates.
Solubility
Soluble in water, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
Chemical Intermediates.
Solubility
Soluble in water, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
Specifications
Specifications
| Melting Point | 66°C to 68°C |
| Density | 0.969 |
| Boiling Point | 198°C to 200°C |
| Flash Point | 93°C (199°F) |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 1720305 |
| Solubility Information | Soluble in water,and methanol. |
| Formula Weight | 100.12 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 208-782-7 |
| RTECSNumber | GQ5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction