Alfa Aesar™ 3,3-Dimethylbutyraldehyde, 97%

Manufacturer: Alfa Aesar™ H55522XF

Quantity 1mL

 View more versions of this product

Catalog No. AAH55522XF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2987-16-8
    Molecular Formula C6H12O
    Linear Formula (CH3)3CCH2CHO
    Formula Weight 100.16
    MDL Number MFCD00042807
    UN Number UN1989
    Beilstein 1735829
    Synonym 3,3-Dimethylbutanal
    Chemical Name or Material 3,3-Dimethylbutyraldehyde

    Specifications

    Formula Weight 100.16
    Linear Formula (CH3)3CCH2CHO
    MDL Number MFCD00042807
    UN Number UN1989
    Beilstein 1735829
    Flash Point 13°C (55°F)
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
    Molecular Formula C6H12O
    Boiling Point 104° to 106°C
    CAS 2987-16-8
    Quantity 1mL
    Synonym 3,3-Dimethylbutanal
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3,3-Dimethylbutyraldehyde
    Assay 97%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit