Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylbutyraldehyde

Catalog No. ac257260010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 2987-16-8 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2987-16-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00042807
InChI Key LTNUSYNQZJZUSY-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyraldehyde, 3,3-dimethyl-butyraldehyde, butanal, 3,3-dimethyl, unii-cam6hd7jki, tert-butylacetaldehyde, 3,3-dimethyl-butanal, cam6hd7jki, neohexal, pubchem3076, 3,3-dimethylbutyraldehyd
PubChem CID 76335
IUPAC Name 3,3-dimethylbutanal
SMILES CC(C)(C)CC=O

Specifications

Melting Point -24°C
Color Colorless
Density 0.7980g/mL
Boiling Point 104°C to 106°C
Flash Point 13°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Linear Formula (CH3)3CCH2CHO
Quantity 1 g
Specific Gravity 0.798
Solubility Information Solubility in water: 7.3g/L (20°C)
Viscosity 0.98 mPa.s (20°C)
Formula Weight 100.16
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylbutyraldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-054-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.