Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylglutaric acid, 98%

Catalog No. AC116240250
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 4839-46-7 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4839-46-7
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00002716
InChI Key DUHQIGLHYXLKAE-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid
PubChem CID 20984
ChEBI CHEBI:68502
IUPAC Name 3,3-dimethylpentanedioic acid
SMILES CC(C)(CC(O)=O)CC(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color White to Beige
Boiling Point 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2C(CH3)2CH2CO2H
Quantity 25 g
Beilstein 02, 684
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether,sparingly soluble in benzene and chloroform,insoluble in low-boiling petroleum fractions
Formula Weight 160.17
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3, 3-Dimethylglutaric acid
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
EINECSNumber 225-425-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.