Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylpiperidine, 95%

Catalog No. AAH2575303
Click to view available options
Quantity:
250 mg
1 g

CAS: 1193-12-0 | C7H15N | 113.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-12-0
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00005995
InChI Key CDODDZJCEADUQQ-UHFFFAOYSA-N
PubChem CID 70942
IUPAC Name 3,3-dimethylpiperidine
SMILES CC1(C)CCCNC1

Specifications

Density 0.81
Boiling Point 134°C to 136°C
Flash Point 26°C (79°F)
Odor Amine-like
Refractive Index 1.4475
Quantity 1 g
UN Number UN2733
Sensitivity Air Sensitive
Synonym piperidine, 3,3-dimethyl, 3,3-dimethyl-piperidine, 3,3-dimethyl piperidine, pubchem10316, acmc-1bo7w, 3,3-dimethylpiperdine
Formula Weight 113.2
Percent Purity 95%
Chemical Name or Material 3,3-Dimethylpiperidine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 214-767-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.