Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenylpropionitrile, Thermo Scientific Chemicals

Catalog No. ac466080050
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2286-54-6
MDL Number MFCD00129747

Specifications

Alpha Vector DIPHENYLPROPIONITRILE 010
Melting Point 86°C to 89°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C15 H13 N
Quantity 5 g
Solubility Information soluble in hot methanol
Molecular Weight (g/mol) 207.27
Formula Weight 207.27
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenylpropionitrile
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.