Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Iminobis(N,N-dimethylpropylamine)

Catalog No. AC466341000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466341000 100 mL
AC466340250 25 mL
AC466345000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC466341000 Supplier Thermo Scientific Chemicals Supplier No. 466341000
Add to Cart

CAS: 6711-48-4 | C10H25N3 | 187.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6711-48-4
Molecular Formula C10H25N3
Molecular Weight (g/mol) 187.33
InChI Key BXYVQNNEFZOBOZ-UHFFFAOYSA-N
IUPAC Name (3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine
SMILES CN(C)CCCNCCCN(C)C

Specifications

Melting Point -78.0°C
Density 0.8410 g/mL
Boiling Point 128.0°C to 131.0°C (27.0 hPa)
Flash Point 98°C
Quantity 100 mL
Specific Gravity 0.841
Formula Weight 187.33
Chemical Name or Material 3,3 '-Iminobis(N,N-dimethylpropylamine)
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Toxic in contact with skin.

Causes severe skin burns and eye damage.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 229-761-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.