Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Pyridyl)acrylic Acid 98.0+%, TCI America™
SDP

Catalog No. P115725G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1126-74-5
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00006410
InChI Key VUVORVXMOLQFMO-ONEGZZNKSA-N
Synonym 3-3-pyridyl acrylic acid, trans-3-3-pyridyl acrylic acid, e-3-pyridin-3-yl acrylic acid, 3-pyridylacrylic acid, 2e-3-pyridin-3-ylacrylic acid, 2e-3-pyridin-3-yl prop-2-enoic acid, 3-pyridin-3-yl acrylic acid, trans-3-3'-pyridyl acrylic acid, 3-3-pyridinyl acrylic acid, 3-pyridinyl-acrylic acid
PubChem CID 776396
IUPAC Name (E)-3-pyridin-3-ylprop-2-enoic acid
SMILES C1=CC(=CN=C1)C=CC(=O)O

Specifications

Melting Point 234°C
Color White
Quantity 25 g
Formula Weight 149.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3-Pyridyl)acrylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.