Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Pyridyl)benzeneboronic acid, 98%

Catalog No. AAH6473903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6473903 1 g
AAH6473906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6473903 Supplier Thermo Scientific Chemicals Supplier No. H6473903

CAS: 351422-72-5 | C11H10BNO2 | 199.02 g/mol

It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351422-72-5
Molecular Formula C11H10BNO2
Molecular Weight (g/mol) 199.02
MDL Number MFCD16658918
InChI Key SSVQXHWHJJERAP-UHFFFAOYSA-N
Synonym 3-pyridin-3-yl phenyl boronic acid, 3-3-pyridyl benzeneboronic acid, 3-pyridin-3-yl phenylboronic acid, 3-3-pyridinyl phenylboronic acid, 3-3-pyridinyl phenyl boronic acid, boronic acid, 3-3-pyridinyl phenyl, 3-3-pyridyl phenylboronic acid, 3-pyridin-3-yl phenyl boronic acid pinacol ester
PubChem CID 58526507
SMILES OB(O)C1=CC(=CC=C1)C1=CN=CC=C1

Specifications

Color White
Quantity 1 g
Beilstein 20520996
IUPAC Name (3-pyridin-3-ylphenyl)boronic acid
Formula Weight 199.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(3-Pyridyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.