Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Pyridyl)-D-alanine, 95%

Catalog No. p-7054734
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAH63132MD

Supplier: Thermo Scientific Chemicals H63132MD

Only null left
Add to Cart

CAS: 70702-47-5 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70702-47-5
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00079673
InChI Key DFZVZEMNPGABKO-UHFFFAOYNA-N
Synonym 3-3-pyridyl-d-alanine, d-3-pyridylalanine, d-3-3-pyridyl-alanine, h-3-d-pal-oh, 3'-pyridyl-d-ala, r-2-amino-3-pyridin-3-yl propanoic acid, 2r-2-amino-3-pyridin-3-yl propanoic acid, 2r-2-amino-3-3-pyridyl propanoic acid, 3-aza-d-phenylalanine, 3-aza-l-phenylalanine
PubChem CID 1268144
SMILES NC(CC1=CN=CC=C1)C(O)=O

Specifications

Color White
Beilstein 4351584
IUPAC Name 2-amino-3-(pyridin-3-yl)propanoic acid
Quantity 250 mg
Formula Weight 166.18
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-(3-Pyridyl)-D-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.