Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,3'-Thiodipropionic acid, 98%
Click to view available options
Quantity:
250 g
1000 g
Description
3,3'-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.
Solubility
Soluble in acetone, ethanol. Slightly soluble in water.
Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.
Solubility
Soluble in acetone, ethanol. Slightly soluble in water.
Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | 127°C to 134°C |
Linear Formula | S(CH2CH2CO2H)2 |
Quantity | 1000 g |
Beilstein | 1210299 |
Merck Index | 14,9332 |
Solubility Information | Soluble in acetone,ethanol. Slightly soluble in water. |
Formula Weight | 178.21 |
Percent Purity | 98% |
Chemical Name or Material | 3,3'-Thiodipropionic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 203-841-3 |
RTECSNumber | UF7990000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction