Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylaluminium chloride, 0.9M solution in heptane, AcroSeal™

Catalog No. AC377798000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 1184-58-3 | C2H6AlCl | 92.502 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-58-3, 142-82-5
MDL Number MFCD00000458
InChI Key JGHYBJVUQGTEEB-UHFFFAOYSA-M
Synonym aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution
PubChem CID 79147
IUPAC Name chloro(dimethyl)alumane
SMILES C[Al](C)Cl

Specifications

Color Colorless to Gray
Density 0.7110g/mL
Flash Point −4°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C2H6AlCl
Quantity 800 mL
Specific Gravity 0.711
Merck Index 15, 3249
Molecular Weight (g/mol) 92.502
Formula Weight 92.5
Physical Form Solution
Chemical Name or Material Dimethylaluminium chloride, 0.9M Solution in Heptane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.