Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Triethoxybenzoic acid, 97%, Thermo Scientific™

Catalog No. AC241870500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241870500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC241870500 Supplier Thermo Scientific Chemicals Supplier No. 241870500

Chemical Identifiers

Molecular Formula C13H18O5
Molecular Weight (g/mol) 254.28
MDL Number MFCD00002505
InChI Key YCENSLDYFDZUMO-UHFFFAOYSA-N
Synonym benzoic acid,3,4,5-triethoxy, benzoic acid, 3,4,5-triethoxy, 3,4,5-triethoxy benzoic acid, acmc-1b720
PubChem CID 81432
IUPAC Name 3,4,5-triethoxybenzoic acid
SMILES CCOC1=CC(=CC(=C1OCC)OCC)C(=O)O

Specifications

CAS Min % 96
CAS Max % 100
Melting Point 109.0°C to 112.0°C
Color Beige
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula (C2H5O)3C6H2COOH
Quantity 50 g
Formula Weight 254.28
Percent Purity 0.97
Physical Form Amorphous Powder
Chemical Name or Material 3, 4, 5-Triethoxybenzoic acid, 0.97
Show More Show Less
EINECSNumber 230-192-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.