Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trifluorophenol 98.0+%, TCI America™
SDP

Catalog No. T229925G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 99627-05-1
Molecular Formula C6H3F3O
Molecular Weight (g/mol) 148.084
MDL Number MFCD00042427
InChI Key ZRTWIJKGTUGZJY-UHFFFAOYSA-N
PubChem CID 2777943
IUPAC Name 3,4,5-trifluorophenol
SMILES C1=C(C=C(C(=C1F)F)F)O

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 177°C
Quantity 25 g
UN Number 3077
Synonym phenol, 3,4,5-trifluoro, 3,4,5-trifluoro-phenol, pubchem2316, 3,4,5-trifluoropheno, acmc-209sdi, ksc486m6r, 3 , 4 , 5-trifluorophenol, phenol, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenol
Formula Weight 148.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trifluorophenol
EINECSNumber (3)-4667
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.