Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trifluorophenylboronic acid, 95+%, Thermo Scientific™

Catalog No. p-3779770
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143418-49-9
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.901
MDL Number MFCD02093069
InChI Key UHDDEIOYXFXNNJ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj
PubChem CID 2734671
IUPAC Name (3,4,5-trifluorophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)F)F)F)(O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 175.9
Percent Purity ≥95.0%
Physical Form Powder
Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.