Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,9,10-Perylenetetracarboxylic diimide

Catalog No. AA4409814
Click to view available options
Quantity:
5 g
25 g

Catalog No. AA4409814

Supplier: Thermo Scientific Chemicals 04409814

Only null left
Add to Cart

CAS: 81-33-4 | C24H10N2O4 | 390.354 g/mol

Used in chemical research. PTCDI is used extensively as an industrial pigment. It is used as tunable laser dye, light-harvesting material, transistor, molecular switched, solar cell, and optoelectronic device. Planar perylene- and naphthalene-based diimide linkers can be employed to tether the Watson- Crick and the Hoogsteen strands of a DNA triplex, thus providing conjugates capable of targeting singlestranded nucleic acids with the formation of hairpin triplexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-33-4
Molecular Formula C24H10N2O4
Molecular Weight (g/mol) 390.354
MDL Number MFCD00024144
InChI Key KJOLVZJFMDVPGB-UHFFFAOYSA-N
Synonym perylimid, 3,4,9,10-perylenetetracarboxylic diimide, perylenediimide, 3,4,9,10-perylenetetracarboxylic acid diimide, perylene-3,4:9,10-tetracarboxydiimide, unii-63nli8842l, anthra 2,1,9-def:6,5,10-d'e'f' diisoquinoline-1,3,8,10 2h,9h-tetrone, anthra 2,1,9-def:6,5,10-d'e'f' diisoquinoline-1,3,8,10 2h,9h-tetraone, isoquino 4',5',6':6,5,10 anthra 2,1,9-def isoquinoline-1,3,8,10 2h,9h-tetrone
PubChem CID 66475
ChEBI CHEBI:52753
SMILES C1=CC2=C3C(=CC=C4C3=C1C5=C6C4=CC=C7C6=C(C=C5)C(=O)NC7=O)C(=O)NC2=O

Specifications

Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 390.35
Physical Form Powder
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic diimide
EINECSNumber 201-344-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.