Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Chlorobenzoyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. C1270100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1270100G 100 g
C127025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1270100G Supplier TCI America Supplier No. C1270100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3984-34-7
Molecular Formula C10H9ClO3
Molecular Weight (g/mol) 212.63
MDL Number MFCD00002794
InChI Key AHVASTJJVAYFPY-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl-4-oxobutanoic acid, 3-4-chlorobenzoyl propionic acid, 4-4-chloro-phenyl-4-oxo-butyric acid, 3-4-chlorobenzoyl propanoic acid, 3-p-chlorobenzoyl propionic acid, 4-4-chlorophenyl-4-oxobutanoicacid, maybridge1_006214, ncistruc1_000312, ncistruc2_000239, asischem d51605
PubChem CID 77604
IUPAC Name 4-(4-chlorophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 100 g
Formula Weight 212.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Chlorobenzoyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.