Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Chlorophenoxy)benzaldehyde 98%, Thermo Scientific™

Catalog No. AC187350100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC187350100 10 g
AC187350500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC187350100 Supplier Thermo Scientific Chemicals Supplier No. 187350100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69770-20-3
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.663
InChI Key MYQFRCYBOOWGJQ-UHFFFAOYSA-N
PubChem CID 616015
IUPAC Name 3-(4-chlorophenoxy)benzaldehyde
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)Cl)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.6070 to 1.6090 (20°C, 589nm)
Linear Formula ClC6H4OC6H4CHO
Quantity 10 g
Synonym 3-4-chlorophenoxy benzaldehyde, m-p-chlorophenoxy benzaldehyde, benzaldehyde,3-4-chlorophenoxy, benzaldehyde, 3-4-chlorophenoxy, acmc-1b8o6, 3-4-chlorophenoxy-benzaldehyde, 3-4-chloranylphenoxy benzaldehyde, 3-4-chloro-phenoxy-benzaldehyde, 3-4-chlorophenoxy benzaldehyde #
Formula Weight 232.67
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 3-(4-Chlorophenoxy)benzaldehyde, 98%
Show More Show Less
EINECSNumber 274-108-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.