Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dehydro-L-proline 99.0+%, TCI America™
SDP

Catalog No. D38251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38251G 1 g
D3825100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38251G Supplier TCI America Supplier No. D38251G

Chemical Identifiers

CAS 4043-88-3
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00065964
InChI Key OMGHIGVFLOPEHJ-BYPYZUCNSA-N
Synonym (S)-3-Pyrroline-2-carboxylic Acid
PubChem CID 94284
IUPAC Name (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid
SMILES OC(=O)[C@H]1NCC=C1

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 1 g
Formula Weight 113.12
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dehydro-L-proline
RTECSNumber UX9371345
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.