Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diaminobenzhydrazide, 98%, Thermo Scientific Chemicals

Catalog No. AAL0526303
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103956-09-8
Molecular Formula C7H10N4O
Molecular Weight (g/mol) 166.184
MDL Number MFCD00017069
InChI Key YGCXQTYRSKMILM-UHFFFAOYSA-N
Synonym 3,4-diaminobenzhydrazide, 4-???benzene-1,2-diamine, acmc-20anfu, 3,4-bis azanyl benzohydrazide, benzoic acid,3,4-diamino-,hydrazide, benzoic acid,3,4-diamino-, hydrazide
PubChem CID 4530104
IUPAC Name 3,4-diaminobenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)N)N

Specifications

Melting Point 148°C to 150°C
Quantity 1 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3,4-Diaminobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.