Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diaminobenzophenone 98.0+%, TCI America™
SDP

Catalog No. D1950100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1950100G 100 g
D195025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1950100G Supplier TCI America Supplier No. D1950100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39070-63-8
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00007727
InChI Key RXCOGDYOZQGGMK-UHFFFAOYSA-N
Synonym 3,4-diaminobenzophenone, 3,4-diaminophenyl phenyl methanone, 3,4-diaminophenyl phenylmethanone, 4-benzoylbenzene-1,2-diamine, 2-amino-4-benzoylaniline, 4-benzoyl-o-phenylenediamine, 3,4-diaminophenyl phenyl ketone, 3,4-diaminophenyl-phenylmethanone, methanone, 3,4-diaminophenyl phenyl, pubchem3382
PubChem CID 135520
IUPAC Name (3,4-diaminophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)N)N

Specifications

Melting Point 118°C
Quantity 100 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diaminobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.