Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4'-Diaminodiphenylmethane 98.0+%, TCI America™
SDP

Catalog No. D168425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D168425G 25 g
D16841G 1 g
D16845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D168425G Supplier TCI America Supplier No. D168425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19430-83-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD00014778
InChI Key FGWQCROGAHMWSU-UHFFFAOYSA-N
PubChem CID 146674
IUPAC Name 3-[(4-aminophenyl)methyl]aniline
SMILES C1=CC(=CC(=C1)N)CC2=CC=C(C=C2)N

Specifications

Melting Point 86°C
Quantity 25 g
Synonym 3,4′-Methylenedianiline
Formula Weight 198.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4′-Diaminodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.