Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diaminotoluene, 97%

Catalog No. AC112435000
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle

CAS: 496-72-0 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-72-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007728
InChI Key DGRGLKZMKWPMOH-UHFFFAOYSA-N
Synonym 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene
PubChem CID 10332
IUPAC Name 4-methylbenzene-1,2-diamine
SMILES CC1=CC(=C(C=C1)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 90.0°C
Color Brown to Brown-Gray
Boiling Point 265.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 500 g
Beilstein 13, 148
Solubility Information Solubility in water: 16g/L (20°C).
Formula Weight 122.17
Percent Purity 97%
Physical Form Crystalline Chunks or Flakes
Chemical Name or Material 3, 4-Diaminotoluene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y
EINECSNumber 207-826-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.