Alfa Aesar™ 3,4-Dibromothiophene, 98+%

Manufacturer: Alfa Aesar™ A1051206

Quantity 5g

 View more versions of this product

Catalog No. AAA1051206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3141-26-2
    Molecular Formula C4H2Br2S
    Formula Weight 241.94
    MDL Number MFCD00005465
    Beilstein 107642
    Chemical Name or Material 3,4-Dibromothiophene

    Specifications

    Formula Weight 241.94
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Molecular Formula C4H2Br2S
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    MDL Number MFCD00005465
    Beilstein 107642
    Boiling Point 221° to 222°C
    CAS 3141-26-2
    Melting Point 4° to 5°C
    Flash Point >110°C (230°F)
    Health Hazard 1 Warning
    Quantity 5g
    Chemical Name or Material 3,4-Dibromothiophene
    Assay ≥98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit