Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzaldehyde, 97%

Catalog No. p-7027722
Click to view available options
Quantity:
5 g
25 g

CAS: 6287-38-3 | C7H4Cl2O | 175.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6287-38-3
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003351
InChI Key ZWUSBSHBFFPRNE-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dichloro, unii-i6z632wsj5, ccris 6015, pubchem8230, 3,4-dichlorobenzaldehye, 3,4-dichlorobenzaldhyde, 3,4 dichlorobenzaldehyde, 3,4-dichlorobenzaidehyde, 4,3-dichlorobenzaldehyde, dsstox_cid_4971
PubChem CID 22710
IUPAC Name 3,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Cl)Cl

Specifications

Melting Point 41°C to 44°C
Boiling Point 247°C to 248°C
Flash Point 135°C (275°F)
Quantity 5 g
UN Number UN1759
Beilstein 636379
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzaldehyde
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 228-520-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.