Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 11.0°C to 13.0°C |
| Density | 0.9820g/mL |
| Boiling Point | 170.0°C |
| Flash Point | 32°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 91% min. (endo-Dicyclopentadiene) (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5100 to 1.5120 |
| Quantity | 2.5 L |
| Beilstein | 05,II,391 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed. Fatal if inhaled. Causes skin irritation. Toxic to aquatic life with long lasting effects. Highly flammable |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Co |
| EINECSNumber | 201-052-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction