Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diethoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™
SDP

Catalog No. D325325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D325325G 25 g
D32535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D325325G Supplier TCI America Supplier No. D325325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5231-87-8
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
InChI Key DFSFLZCLKYZYRD-UHFFFAOYSA-N
Synonym 3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn
PubChem CID 123228
IUPAC Name 3,4-diethoxycyclobut-3-ene-1,2-dione
SMILES CCOC1=C(C(=O)C1=O)OCC

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
Formula Weight 170.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Diethoxy-3-cyclobutene-1,2-dione
RTECSNumber GU1772466
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.