Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorobenzoic acid, 98%

Catalog No. AAA1587606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1587606 5 g
AAA1587603 1 g
AAA1587614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AAA1587606 Supplier Thermo Scientific Chemicals Supplier No. A1587606
Add to Cart
This item is not returnable. View return policy

CAS: 455-86-7 | C7H4F2O2 | 158.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-86-7
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00011672
InChI Key FPENCTDAQQQKNY-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721
PubChem CID 99166
IUPAC Name 3,4-difluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)F

Specifications

Melting Point 121°C to 124°C
Odor Odorless
Quantity 5 g
Beilstein 2085848
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-249-6
TSCA Yes

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.