Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-difluorobenzyl Chloride, 98%

Catalog No. p-3211785
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 698-80-6 | C7H5ClF2 | 162.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-80-6
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.56
MDL Number MFCD00236218
InChI Key VTGRVYJPIVZZGS-UHFFFAOYSA-N
Synonym 3,4-difluorobenzyl chloride, 4-chloromethyl-1,2-difluorobenzene, 3,4-difluorobenzylchloride, benzene, 4-chloromethyl-1,2-difluoro, 1-chloromethyl-3,4-difluorobenzene, alpha-chloro-3,4-difluorotoluene, 3, 4-difluorobenzyl chloride, pubchem7357, 4-chloromethyl-1,2-difluoro-benzene
PubChem CID 2736954
IUPAC Name 4-(chloromethyl)-1,2-difluorobenzene
SMILES FC1=CC=C(CCl)C=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Linear Formula ClCH2C6H3F2
Quantity 5 g
Formula Weight 162.57
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorobenzyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.