Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluoroiodobenzene, 99%, Thermo Scientific™

Catalog No. ac233180250
Click to view available options
:
25g; Glass bottle

Specifications

CAS 64248-58-4
CAS Min % 98.5
CAS Max % 100.0
Density 1.9900g/mL
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H3F2I
Refractive Index 1.5570 to 1.559
Linear Formula IC6H3F2
Quantity 25g
MDL Number MFCD00044630
Specific Gravity 1.99
Synonym 3,4-difluoroiodobenzene, 1,2-difluoro-4-iodo-benzene, benzene, 1,2-difluoro-4-iodo, 3,4-difluoro iodobenzene, pubchem3461, 3,4-difluoro-iodobenzene, 3,4-difluorophenyl iodide, 3,4-difluoro-1-iodobenzene, 1-iodo-3,4-difluoro-benzene
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
InChI Key KSASJELKLBIMSG-UHFFFAOYSA-N
SMILES FC1=C(F)C=C(I)C=C1
IUPAC Name 1,2-difluoro-4-iodobenzene
Molecular Weight (g/mol) 239.99
PubChem CID 2724444
Formula Weight 239.99
Percent Purity 99%
Chemical Name or Material 3, 4-Difluoroiodobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.