Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorophenyl Methyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. D45261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45261G 1 g
D45265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45261G Supplier TCI America Supplier No. D45261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 424792-57-4
Molecular Formula C7H6F2O2S
Molecular Weight (g/mol) 192.18
MDL Number MFCD04037934
InChI Key WMBJGJXMKCOHGG-UHFFFAOYSA-N
PubChem CID 2782743
IUPAC Name 1,2-difluoro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC(=C(C=C1)F)F

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
Formula Weight 192.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluorophenyl Methyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.