Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2-methoxy-2H-pyran 97.0+%, TCI America™
SDP

Catalog No. D155425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D155425ML 25 mL
D1554500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D155425ML Supplier TCI America Supplier No. D155425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4454-05-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00006560
InChI Key XCYWUZHUTJDTGS-UHFFFAOYNA-N
Synonym 3,4-dihydro-2-methoxy-2h-pyran, 2h-pyran, 3,4-dihydro-2-methoxy, 2,3-dihydro-2-methoxy 4h pyran, 2-methoxy-2,3-dihydro-4h-pyran, acmc-1ahgh, dsstox_cid_7570, dsstox_rid_78508, dsstox_gsid_27570, ksc496o2j
PubChem CID 91538
IUPAC Name 2-methoxy-3,4-dihydro-2H-pyran
SMILES COC1CCC=CO1

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 mL
UN Number 1993
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 3,4-Dihydro-2-methoxy-2H-pyran
EINECSNumber (5)-5581
RTECSNumber UP7900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.