Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine, 97%, Thermo Scientific™

Catalog No. CC60830DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC60830DA 1 g Amber Glass Bottle
CC60830EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC60830DA Supplier Thermo Scientific Chemicals Supplier No. CC60830DA
Add to Cart

Chemical Identifiers

CAS 20348-23-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD09025907
InChI Key QQVXDMFULJVZLA-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine, 3,4-dihydro-2h-pyrido 3,2-b-1,4-oxazine, 2h,3h,4h-pyrido 3,2-b 1,4 oxazine, 2h-pyrido 3,2-b-1,4-oxazine, 3,4-dihydro, dihydropyridoboxazine, pubchem18891, 2h,3h,4h-pyridino 2,3-e 1,4-oxazine, 2h-pyrido 3,2-b-1,4-oxazine,3,4-dihydro
PubChem CID 13196538
IUPAC Name 3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
SMILES C1COC2=C(N1)N=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-pyrido[3,2-b][1,4]oxazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.