Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-7-hydroxy-2(1H)-quinolinone 98.0+%, TCI America™
SDP

Catalog No. D359925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D359925G 25 g
D35995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D359925G Supplier TCI America Supplier No. D359925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22246-18-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD06410891
InChI Key LKLSFDWYIBUGNT-UHFFFAOYSA-N
Synonym 3,4-dihydro-7-hydroxy-2 1h-quinolinone, 7-hydroxy-3,4-dihydroquinolin-2 1h-one, 7-hydroxy-3,4-dihydro-2 1h-quinolinone, 3,4-dihydro-7-hydroxycarbostyril, 7-hydroxy-3,4-dihydro-quinolin-2-one, 7-hydroxy-3,4-dihydrocarbostyril, 7-hydroxy-3,4-dihydro carbostyril, 7-hydroxy-1,2,3,4-tetrahydroquinolin-2-one, 2 1h-quinolinone, 3,4-dihydro-7-hydroxy, 7-hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline
PubChem CID 2785758
IUPAC Name 7-hydroxy-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)O

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 25 g
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydro-7-hydroxy-2(1H)-quinolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.