Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxybenzhydrazide, 97%

Catalog No. AAB2174614
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2174614

Supplier: Thermo Scientific Chemicals B2174614

Only null left
Add to Cart

CAS: 39635-11-5 | C7H8N2O3 | 168.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39635-11-5
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00017064
InChI Key WGXWEXNJRZMIPT-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzhydrazide, 3,4-dihydroxy-benzoic acid hydrazide, 4-???benzene-1,2-diol, protocatechuhydrazide, acmc-1ad2t, 3,4-dihydroxy-benzohydrazide, 3,4-bis oxidanyl benzohydrazide, 3,4-dihydroxybenzoic acid hydrazide, 3,4-dihydroxy-benzoesaeure-hydrazid, 3,4-dihydroxybenzoic acid, hydrazide
PubChem CID 586300
IUPAC Name 3,4-dihydroxybenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)O)O

Specifications

Melting Point ∼265°C (decomposition)
Quantity 25 g
Beilstein 2642968
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 3,4-Dihydroxybenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-550-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.