Learn More
3,4-Dihydroxybenzoic acid, 97%
Description
3,4-Dihydroxybenzoic acid is a metabolite of polyphenols such as phloretin and quercetin. It is also a phenolic acid antioxidant present in fruits, vegetables and nuts. Further, it is found to be an efficacious chemopreventive agent in several carcinogenesis models. In addition to this, it acts as an anti-inflammatory agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,4-Dihydroxybenzoic acid is a metabolite of polyphenols such as phloretin and quercetin. It is also a phenolic acid antioxidant present in fruits, vegetables and nuts. Further, it is found to be an efficacious chemopreventive agent in several carcinogenesis models. In addition to this, it acts as an anti-inflammatory agent.
Solubility
Soluble in hot alcohol and ether. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.

Specifications
Specifications
Melting Point | ∼202°C (decomposition) |
Quantity | 100 g |
Beilstein | 1448841 |
Merck Index | 14,7894 |
Solubility Information | Soluble in hot alcohol and ether. Slightly soluble in water. |
Formula Weight | 154.12 |
Percent Purity | 97% |
Chemical Name or Material | 3,4-Dihydroxybenzoic acid |
Safety and Handling
Hazard Category | H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 202-760-0 |
RTECSNumber | UL0560000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.