Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer

Catalog No. p-4258958
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 331-39-5 | C9H8O4 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
InChI Key QAIPRVGONGVQAS-DUXPYHPUSA-N
Synonym caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
PubChem CID 689043
ChEBI CHEBI:16433
IUPAC Name (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 194.0°C to 198.0°C
Color Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (HO)2C6H3CH=CHCO2H
Quantity 5 g
Beilstein 10,436
Merck Index 15,1638
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in cold alcohol
Formula Weight 180.16
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 3, 4-Dihydroxycinnamic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 206-361-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.