Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxycinnamic acid, predominantly trans, 98+%

Catalog No. p-7022947
Click to view available options
Quantity:
10 g
50 g

CAS: 331-39-5 | C9H8O4 | 180.159 g/mol

3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

Solubility
Soluble in DMSO (40 mg/mol), ethanol (25 mg/mL warm), DMF (∼7 mg/mL), PBS, PH 7.2 (∼0.65 mg/mol), hot water, ethyl acetate, Chloroform Hexane, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00004392
InChI Key QAIPRVGONGVQAS-DUXPYHPUSA-N
Synonym caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
PubChem CID 689043
ChEBI CHEBI:16433
IUPAC Name (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)O)O

Specifications

Melting Point ∼202°C (decomposition)
Quantity 10 g
Beilstein 2210883
Merck Index 14,1635
Solubility Information Soluble in DMSO (40mg/mol),ethanol (25mg/mL warm),DMF (∽7mg/mL),PBS,PH 7.2 (∽0.65mg/mol),hot water,ethyl acetate,Chloroform Hexane,and methanol.
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material 3,4-Dihydroxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335-H351-H361
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 206-361-2
RTECSNumber GD8950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.