Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dihydroxyflavone 97.0+%, TCI America™
SDP

Catalog No. D48841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48841G 1 g
D4884200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48841G Supplier TCI America Supplier No. D48841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4143-64-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00017600
InChI Key SRNPMQHYWVKBAV-UHFFFAOYSA-N
Synonym 3',4'-dihydroxyflavone, 2-3,4-dihydroxyphenyl-4h-chromen-4-one, 2-3,4-dihydroxyphenyl chromen-4-one, unii-koh101s66v, 3',4'-dihydroxy flavone, 3,4-dihydroxyflavone, 4h-1-benzopyran-4-one, 2-3,4-dihydroxyphenyl, dihydroxyflavone, 3',4', 4hlm, pubchem9842
PubChem CID 145726
IUPAC Name 2-(3,4-dihydroxyphenyl)chromen-4-one
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C3=CC(=C(C=C3)O)O

Specifications

Melting Point 248°C
Quantity 1 g
Formula Weight 254.24
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dihydroxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.