Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydroxyphenylacetic acid, 98%

Catalog No. p-3778309
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 102-32-9 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-32-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00004338
InChI Key CFFZDZCDUFSOFZ-UHFFFAOYSA-N
Synonym 3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate
PubChem CID 547
ChEBI CHEBI:41941
IUPAC Name 2-(3,4-dihydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 126°C to 130°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CH2CO2H
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 168.15
Percent Purity 98%
Physical Form Powder, Crystals or Lumps
Chemical Name or Material 3, 4-Dihydroxyphenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 203-024-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.