Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™
SDP

Catalog No. D209925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D209925G 25 g
D20991G 1 g
D20995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D209925G Supplier TCI America Supplier No. D209925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5222-73-1
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00101316
InChI Key SZBNZTGCAMLMJY-UHFFFAOYSA-N
PubChem CID 123227
IUPAC Name 3,4-dimethoxycyclobut-3-ene-1,2-dione
SMILES COC1=C(C(=O)C1=O)OC

Specifications

Melting Point 56°C
Color White
Quantity 25 g
Synonym 3,4-dimethoxy-3-cyclobutene-1,2-dione, dimethyl squarate, squaric acid dimethyl ester, 3-cyclobutene-1,2-dione, 3,4-dimethoxy, 3,4-dimethoxy-cyclobut-3-ene-1,2-dione, dimethoxycyclobut-3-ene-1,2-dione, 3-cyclobutene-1,2-dione,3,4-dimethoxy, 3,4-dimethoxycyclobut-3-en-1,2-dion, dimethoxycyclobutenedione, pubchem13709
Formula Weight 142.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxy-3-cyclobutene-1,2-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.