Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzoic acid, 99+%

Catalog No. p-3757729
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115450250 25 g Glass bottle
AC115451000 100 g Glass bottle
AC115455000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC115450250 Supplier Thermo Scientific Chemicals Supplier No. 115450250
Only null left
Add to Cart

CAS: 93-07-2 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-07-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002500
InChI Key DAUAQNGYDSHRET-UHFFFAOYSA-N
Synonym veratric acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, veratrylic acid, 3,4-dimethylprotocatechuic acid, veratrumenoic acid, 3,4-dimethoxy-benzoic acid, unii-1yy04e7rr4, 3,4-dimethoxybenzoes
PubChem CID 7121
ChEBI CHEBI:296881
IUPAC Name 3,4-dimethoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)OC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 179°C to 182°C
Color Beige to White or Red
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CO2H
Quantity 25 g
Beilstein 10, I, 188
Merck Index 15, 10147
Solubility Information Solubility in water: 0.5g/L (14°C). Other solubilities: readily soluble in alcohol,soluble in ether
Formula Weight 182.18
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethoxybenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 202-215-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.