Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzyl alcohol, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
93-03-8
C9H12O3
168.19
OEGPRYNGFWGMMV-UHFFFAOYSA-N
3,4-dimethoxybenzyl alcohol,veratryl alcohol,3,4-dimethoxyphenyl methanol,benzenemethanol, 3,4-dimethoxy,3,4-dimethoxyphenylmethyl alcohol,veratrole alcohol,3,4-dimethoxybenzenemethanol,unii-mb4t4a711h,3,4-dimethoxyphenyl-methanol
7118
CHEBI:62150
(3,4-dimethoxyphenyl)methanol
COC1=C(C=C(C=C1)CO)OC
93-03-8
C9H12O3
168.19
OEGPRYNGFWGMMV-UHFFFAOYSA-N
3,4-dimethoxybenzyl alcohol,veratryl alcohol,3,4-dimethoxyphenyl methanol,benzenemethanol, 3,4-dimethoxy,3,4-dimethoxyphenylmethyl alcohol,veratrole alcohol,3,4-dimethoxybenzenemethanol,unii-mb4t4a711h,3,4-dimethoxyphenyl-methanol
7118
CHEBI:62150
(3,4-dimethoxyphenyl)methanol
COC1=C(C=C(C=C1)CO)OC
Color Colorless to Yellow
Flash Point >112°C
Melting Point 22.0°C
Quantity 100g
Boiling Point 296.0°C to 297.0°C (732.0mmHg)
Density 1.1500g/mL
Specific Gravity 1.15
Refractive Index 1.5510 to 1.5540
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (CH3O)2C6H3CH2OH
Beilstein 06,1113
Solubility Information Solubility in water: miscible
Formula Weight 168.19
Physical Form Oily Liquid
Percent Purity 96%
Chemical Name or Material 3, 4-Dimethoxybenzyl alcohol
Show More Show Less

GHS P Statement Harmful if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Warning