Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzyl alcohol, 97%

Catalog No. AAA1339622
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 93-03-8 | C9H12O3 | 168.192 g/mol

3,4-Dimethoxybenzyl alcohol used as the fuel of the microbial fuel cell (MFC) to generate power. Veratrole alcohol is the raw material for the synthesis of cyclotriveratrylene which is used in host-guest chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzyl alcohol used as the fuel of the microbial fuel cell (MFC) to generate power. Veratrole alcohol is the raw material for the synthesis of cyclotriveratrylene which is used in host-guest chemistry.

Solubility
Fully miscible with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-03-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004638
InChI Key OEGPRYNGFWGMMV-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
PubChem CID 7118
ChEBI CHEBI:62150
IUPAC Name (3,4-dimethoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)OC

Specifications

Melting Point ∼22°C
Density 1.176
Boiling Point 297°C to 298°C
Flash Point >110°C (230°F)
Refractive Index 1.553
Quantity 100 g
Beilstein 639388
Solubility Information Fully miscible with water.
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzyl alcohol
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-212-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.