Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzylamine 97.0+%, TCI America™
SDP

Catalog No. D402425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D402425G 25 g
D40245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D402425G Supplier TCI America Supplier No. D402425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5763-61-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00008116
InChI Key DIVNUTGTTIRPQA-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
PubChem CID 79832
IUPAC Name 1-(3,4-dimethoxyphenyl)methanamine
SMILES COC1=CC=C(CN)C=C1OC

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 g
UN Number 2735
Formula Weight 167.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethoxybenzylamine
RTECSNumber YX7355000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.