Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxycinnamonitrile, Mixture of cis and trans 97%, Thermo Scientific™

Catalog No. AC163360100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163360100 10 g
AC163360500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163360100 Supplier Thermo Scientific Chemicals Supplier No. 163360100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6443-72-7
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001931
InChI Key WGPTVEZJYRFEGT-ONEGZZNKSA-N
Synonym 3,4-dimethoxycinnamonitrile, 3-3,4-dimethoxyphenyl acrylonitrile, 2e-3-3,4-dimethoxyphenyl acrylonitrile, 2e-3-3,4-dimethoxyphenyl prop-2-enenitrile, e-3-3,4-dimethoxyphenyl acrylonitrile, e-3-3,4-dimethoxyphenyl prop-2-enenitrile, .beta.-3,4-dimethoxyphenyl acrylonitrile, cinnamonitrile, 3,4-dimethoxy, 2-propenenitrile, 3-3,4-dimethoxyphenyl, cambridge id 5107337
PubChem CID 1550958
IUPAC Name (2E)-3-(3,4-dimethoxyphenyl)prop-2-enenitrile
SMILES COC1=CC=C(\C=C\C#N)C=C1OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Linear Formula (CH3O)2C6H3CH=CHCN
Quantity 10 g
Formula Weight 189.21
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethoxycinnamonitrile, mixture of cis and trans, 97%
Hazard Category GHS Signal Word: Warning
EINECSNumber 229-240-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.